Pipette tips
Type E, 10 µl, -, Transparent, Eppendorf Reference, Research, mLine Mechanical, Proline Mechanical, eLine Electronic, Tip stack pack, 96/tray, 576/stack pack, 2304/case
Catalog # SARS70.3010.100
Supplier: SARSTEDT
Jump to:
Product Details & Documents
These high quality tips are suitable for the most common pipettes.
- Sterile tips are DNase/RNase- and ATP-free, and non pyrogenic
- Available loose in bags, packed in racks or in Tip StackPack reload systems
Tip StackPack: Tips are preloaded in reusable, stacked 96-tip trays. The trays are designed to safely cover each individual 96-tip layer, shielding the preloaded tips in the stack like a protective screen.
Specifications
- Packed:Tip stack pack, 96/tray, 576/stack pack, 2304/case
- Sterile:-
- Volume:10 µl
- Seq:16
- Tip style:Type E
- Tip colour:Transparent
- For use with:Eppendorf Reference, Research, mLine Mechanical, Proline Mechanical, eLine Electronic