Orders LinkContactUsLinkComponent
Pipette tips
Pipette tips
Type D, 10 µl, -, Transparent, Eppendorf Reference, Research and Research Plus, 96/box, 1920/case
Catalog # SARS70.1115.100
Pipette tips
Catalog # SARS70.1115.100
Supplier:  
Pipette tips
Type D, 10 µl, -, Transparent, Eppendorf Reference, Research and Research Plus, 96/box, 1920/case
Discontinued
Product Details & Documents
These high quality tips are suitable for the most common pipettes.
  • Sterile tips are DNase/RNase- and ATP-free, and non pyrogenic
  • Available loose in bags, packed in racks or in Tip StackPack reload systems
Tip StackPack: Tips are preloaded in reusable, stacked 96-tip trays. The trays are designed to safely cover each individual 96-tip layer, shielding the preloaded tips in the stack like a protective screen.
Specifications
  • Packed:
    96/box, 1920/case
  • Sterile:
    -
  • Volume:
    10 µl
  • Seq:
    10
  • Tip style:
    Type D
  • Tip colour:
    Transparent
  • For use with:
    Eppendorf Reference, Research and Research Plus