Orders LinkContactUsLinkComponent
Sample vials, 6 ml
Sample vials, 6 ml
6,00 ml, 22×38 mm, Clear, No
Catalog # METR6.2419.007
Supplier:   METROHM
Sample vials, 6 ml
Catalog # METR6.2419.007
Supplier:  METROHM
Sample vials, 6 ml
6,00 ml, 22×38 mm, Clear, No
Price
162,00 €
Log in for availability
1 * 100 items
Product Details & Documents
Specifications
  • Cap included:
    No
  • Ø×H:
    22×38 mm
  • Capacity:
    6,00 ml
  • Colour:
    Clear
  • Seq:
    02