Safety shields
Straight, 900×10×670 mm
113-5031
Protective shield made from acrylic glass to be used as splinter shield and splash guard.
- Strong and stable design
- Removable feet
- Form:Straight
- W×D×H:900×10×670 mm
Protective shield made from acrylic glass to be used as splinter shield and splash guard.