Orders LinkContactUsLinkComponent
Dispenser Boxes
4 chambers, Clear, 290×160×215 mm
  212-9272
Dispenser Boxes
  212-9272
Dispenser Boxes
Dispenser Boxes
4 chambers, Clear, 290×160×215 mm
329,00 €
1 * 1 items
Acrylic, clear.
  • Easy top loading and convenient dispensing through the front flap
  • Clear front allows contents to be checked easily
  • Cover and front flap limit infiltration by dust and other particles
Ideal for storing and dispensing a wide variety of small parts, such as pipette tips, microtubes, disposable syringes, canulas, etc.
  • Colour:
    Clear
  • Description:
    4 chambers
  • Seq:
    03
  • W×D×H:
    290×160×215 mm