Orders LinkContactUsLinkComponent
System Boxes
Transparent, 230×150×130 mm
  216-3496
System Boxes
  216-3496
System Boxes
System Boxes
Transparent, 230×150×130 mm
17,70 €
1 * 1 items
Boxes made of transparent glass-clear polystyrene. Suitable for variable applications.
  • Provides immediate overview about content and quantity
  • Large sight opening
  • High stackability
  • Wide labelling pad
  • Excellent stability
  • Colour:
    Transparent
  • Dimensions:
    230×150×130 mm
  • Seq:
    02