ContactUsLinkComponent
Valve, with luer lock connectors, made of PC, flow-tap, colour blue, swivel LL-nut, pressure tested, male chicks from HDPE (not auto clavable), connection 1 LL w, 1 LL m. 1 * 1 items
  229-7441
Valve, with luer lock connectors, made of PC, flow-tap, colour blue, swivel LL-nut, pressure tested, male chicks from HDPE (not auto clavable), connection 1 LL w, 1 LL m. 1 * 1 items
  229-7441
Frequently Bought Together