ContactUsLinkComponent
Manifold, with luer lock connector, 3 way valve bank, connection 5 LL female, flat design with cross handle, with end caps, pressure -tested, colour transparent, male chicks HDPE (not autoclavable). 1 * 1 items
  229-7446
Manifold, with luer lock connector, 3 way valve bank, connection 5 LL female, flat design with cross handle, with end caps, pressure -tested, colour transparent, male chicks HDPE (not autoclavable). 1 * 1 items
  229-7446
Frequently Bought Together