Manifold, with luer lock connector, 5 way valve bank, connection 7 LL female, flat design with cross handle, with end caps, pressure -tested, colour transparent, male chicks HDPE (not autoclavable). 1 * 1 items
229-7447
Frequently Bought Together