Orders LinkContactUsLinkComponent
Assortment Boxes, PP
Assortment box, with hinged lid and 4×1, 2×1 and 1×1 compartments, Anthracite, 175×140×30 mm
Catalog # SEMA1770
Supplier:   SEMADENI
Assortment Boxes, PP
Catalog # SEMA1770
Supplier:  SEMADENI
Assortment Boxes, PP
Assortment Boxes, PP
Assortment box, with hinged lid and 4×1, 2×1 and 1×1 compartments, Anthracite, 175×140×30 mm
Price
21,70 €
Log in for availability
1 * 1 items
Product Details & Documents
These assortment boxes, made of PP, have a transparent stable hinged lid with lock and tamper-evident seals.
  • Robust and strong design
  • All models can be stacked together
  • Lid is fixed on each tray so that the contents don't mix even if box is turned upside down
Specifications
  • Colour:
    Anthracite
  • Description:
    Assortment box, with hinged lid and 4×1, 2×1 and 1×1 compartments
  • Seq:
    01
  • W×D×H:
    175×140×30 mm