Face shields, Bionic™
Uncoated, Acetate
111-0252
This innovative face shield provides protection from extreme mechanical hazards. It can be worn in combination with safety spectacles or respiratory protection.
- With extended protection for brow and chin
- Locking mechanism for added safety
- Extra soft padding with washable or replaceable sweatband
- Various adjustments for best comfort
Weight: 355 g.
EN 166, EN 169, EN 170, 1 F, B, T, C, 3, 8, 9
: EN 166, EN 169, EN 170, 1 F, B, T, C, 3, 8, 9
- Type:Uncoated
- Material:Acetate