Screw caps for vials with thread, Nalgene®
Closures lined, black, sterile
Catalog # 215-1507
Supplier: Thermo Fisher Scientific
Screw caps for vials with thread, Nalgene®
Catalog # 215-1507
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Specifications
- Type:Closures lined, black, sterile
- Environmental certification:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact
- Cat. No.:215-1507