Orders LinkContactUsLinkComponent
Mops, LoopMop™
Mops, LoopMop™
LoopMop™
Catalog # 129-0241
Supplier:   MICRONOVA
Mops, LoopMop™
Catalog # 129-0241
Supplier:  MICRONOVA
Mops, LoopMop™
Mops, LoopMop™
LoopMop™
Price
67,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Knitted polyester mop for applying disinfectant in pharmaceutical, aseptic processing and medical device applications. Absorbent enough to clean to a dry finish but will leave enough moisture for disinfectants that require an extended drying time. Mop heads can be used directly with the fibreglass/polypropylene handle or attached to the universal handle via the quick disconnect adapter.
  • Can be laundered or autoclaved
  • Low linting
  • No raw edges
Ordering information: All components must be ordered separately.
Specifications
  • Description:
    LoopMop™
Accessories
Product Information
  • Catalog #
    Length
    Material
    Availability
    Price
  • Stainless steel quick disconnect extendable handle
    Stainless steel quick disconnect extendable handle
    129-0551
    965 - 1829 mm
    Stainless steel 304
    Log in for availability
    1 * 1 items - 358,00 €
Product Information
  • Catalog #
    Length
    Material
    Availability
    Price
  • Stainless steel quick disconnect handle
    Stainless steel quick disconnect handle
    129-0062
    155 cm
    Fibreglass
    Log in for availability
    1 * 1 items - 165,00 €
Product Information
  • Catalog #
    Length
    Material
    Availability
    Price
  • Stainless steel quick disconnect handle
    Stainless steel quick disconnect handle
    129-0176
    1295 cm
    Stainless steel 304
    Log in for availability
    1 * 1 items - 159,00 €
Product Information
  • Catalog #
    Availability
    Price
  • MicroMop® Adapter: PP clamp adapter to hold Micromop and PolyGen series of floor mops, attaches to SSU series of handles
    MicroMop® Adapter: PP clamp adapter to hold Micromop and PolyGen series of floor mops, attaches to SSU series of handles
    129-0170
    Log in for availability
    1 * 1 items - 157,00 €