Mops, MicroMop™
Mop head, polyester
Catalog # 129-0234
Supplier: MICRONOVA
Jump to:
Product Details & Documents
MicroMop™ are ideal for quick and efficient cleaning of large surface areas. Suitable for wafer processing, electronics, pharmaceutical, and biotech applications. All mop heads attach to stainless steel universal handle via the orange polypropylene quick disconnect adapter. For less critical applications, choose the fibreglass/polypropylene handle with attached adapter.
- Low-linting, quick drying PVA is ideal for high-tech applications. Can be laundered
- Polyester has good wicking capability, can be autoclaved and used with strong disinfectants
- Lite™ is sturdy latex/rayon blend for controlled environments and cleanroom support areas; mop head can be autoclaved for medical and sterile applications
Ordering information: All components must be ordered separately.
Specifications
- Description:Mop head, polyester
- Seq:03
- Cat. No.:129-0234
- Environmentally Preferable:Waste Reducing
Accessories
Product Information
- Catalog #AvailabilityPrice
- MicroMop® Adapter: PP clamp adapter to hold Micromop and PolyGen series of floor mops, attaches to SSU series of handlesMicroMop® Adapter: PP clamp adapter to hold Micromop and PolyGen series of floor mops, attaches to SSU series of handles129-0170Log in for availability1 * 1 items - 157,00 €