Pipette tips
Gilson type, 100 - 1000 µl, -, Blue, Gilson P500/P1000, Eppendorf, Socorex, Nichiryo, HTL, 10 racks, 96 each
Supplier: DELTALAB
Jump to:
Product Details & Documents
Documents
Specifications
- Packed:10 racks, 96 each
- Sterile:-
- Volume:100 - 1000 µl
- Seq:18
- Tip style:Gilson type
- Tip colour:Blue
- For use with:Gilson P500/P1000, Eppendorf, Socorex, Nichiryo, HTL