Orders LinkContactUsLinkComponent
Colony counters
Colony counters
Colony counter manual with magnifying glass ø 120 mm with lighting and support
Catalog # 710-0108
Colony counters
Catalog # 710-0108
Supplier:  
Colony counters
Colony counters
Colony counter manual with magnifying glass ø 120 mm with lighting and support
Discontinued
Product Details & Documents
Choice of models with a fibre marker with a built-in switch and /or a metal-tipped pen that are attached to the base. The marker supplied with the counter allows marked cells to be added to the screen one by one.
  • Ø 100 or 120 mm biconvex lamp
  • X2 magnification
220 V voltage, 50 Hz, 0,2 A.
Specifications
  • Description:
    Colony counter manual with magnifying glass ø 120 mm with lighting and support
  • Seq:
    02
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Centering Device 100 mm
    Centering Device 100 mm
    710-0111
    Log in for availability
    1 * 1 items - 61,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Centering Device 80 mm
    Centering Device 80 mm
    710-0110
    Log in for availability
    1 * 1 items - 61,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Centering Device 60 mm
    Centering Device 60 mm
    710-0109
    Log in for availability
    1 * 1 items - 61,00 €