Pipette tips
Gilson type, crystal tip, graduated, 0,1 - 10 µl, -, Transparent, Eppendorf, Labsystems, Biohit, Nichiryo, Rainin, Bulk (20×1000)
Catalog # DELT200024
Supplier: DELTALAB
Jump to:
Product Details & Documents
Documents
Specifications
- Packed:Bulk (20×1000)
- Sterile:-
- Volume:0,1 - 10 µl
- Seq:01
- Tip style:Gilson type, crystal tip, graduated
- Tip colour:Transparent
- For use with:Eppendorf, Labsystems, Biohit, Nichiryo, Rainin