Adapters for GL screw threads
Adapters with conical ground joint 14/23, GL 18
Catalog # QVFLHAA6012504
Supplier: QVF LABORTECHNIK
Jump to:
Product Details & Documents
For connecting GL screw threads and conical ground joints.
Specifications
- Thread:GL 18
- Description:Adapters with conical ground joint 14/23
- Seq:01