Screw caps for internal thread sample tubes
Screw caps with silicone washer, Yellow
SIMPT500YOS
These polypropylene screw caps are to be used with T500 series internal thread sample tubes.
- Made of polypropylene
- Cap with silicone washer for leakproof seal
- Long skirt and super-fast thread design for easy seal
- Autoclavable
Caps are available with a silicone washer between the cap and the tube to ensure a positive leakproof seal at all temperatures. The cap features a long skirt and a super-fast thread design allowing it to be removed or screwed on with a single turn.
These silicone washer caps will guarantee a positive leakproof seal at all temperatures.
: Tubes T500 series sold separately.
- Colour:Yellow
- Description:Screw caps with silicone washer