Capillary columns for ASTM method
Columns used for cumene analysis, Capillary, 0,53 mm, 50 m
Catalog # VARICP7428
Supplier: VARIAN
Jump to:
Product Details & Documents
Specifications
- Column Format:Capillary
- Ø int.:0,53 mm
- Description:Columns used for cumene analysis
- Length:50 m