Orders LinkContactUsLinkComponent
Polyoxyl Stearyl Ether, Reference Standards
Polyoxyl Stearyl Ether, Reference Standards
Polyoxyl 10 Stearyl Ether, USP reference standard, Vial, 1 g
Catalog # 1372606.
Supplier:   USP
Polyoxyl Stearyl Ether, Reference Standards
Catalog # 1372606.
Supplier:  USP
CAS Number:  
Polyoxyl Stearyl Ether, Reference Standards
Polyoxyl 10 Stearyl Ether, USP reference standard, Vial, 1 g
CAS Number:  
Price
231,00 €
Log in for availability
1 * 1 g
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Documents
Specifications
Specifications
  • Pack type:
    Vial
  • Pk:
    1 g
  • Standards:
    USP reference standard
  • Description:
    Polyoxyl 10 Stearyl Ether
  • Cat. No.:
    1372606.
  • Description:
    Polyoxyl 10 Stearyl Ether
  • CAS No.:
    9005-00-9