Orders LinkContactUsLinkComponent
Cables for Electrodes
Cable KA19, 4 cords, 6 mm Ø, max. temperature 130 °C
Catalog # 662-2238
Supplier:   SI Analytics
Cables for Electrodes
Catalog # 662-2238
Supplier:  SI Analytics
Cables for Electrodes
Cables for Electrodes
Cable KA19, 4 cords, 6 mm Ø, max. temperature 130 °C
Price
30,00 €
Log in for availability
1 * 1 m
Specifications
  • Description:
    Cable KA19, 4 cords, 6 mm Ø, max. temperature 130 °C
  • Seq:
    04