Order Entry
Slovenia
Orders LinkContactUsLinkComponent
Erlenmeyer flasks with standard ground joints, amber

Erlenmeyer flasks with standard ground joints, amber

250 ml, Amber, 85 mm, 29/32 NS
Catalog # 201-0929
Supplier:   Lenz Laborglas
Erlenmeyer flasks with standard ground joints, amber
Catalog # 201-0929
Supplier:  Lenz Laborglas
Erlenmeyer flasks with standard ground joints, amber
Erlenmeyer flasks with standard ground joints, amber

250 ml, Amber, 85 mm, 29/32 NS

Price
71,00 €
Log in for availability
1 * 1 items
Product Details & Documents

DURAN® borosilicate glass, brown.

  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes

Coloured brown to protect light-sensitive substances. Volume graduations and labelling area.

Documents

Specifications
  • Ext.Ø:
    85 mm
  • Socket size:
    29/32 NS
  • Capacity:
    250 ml
  • Colour:
    Amber
  • Seq:
    09