Order Entry
Slovenia
Orders LinkContactUsLinkComponent
Erlenmeyer flasks with standard ground joints, amber

Erlenmeyer flasks with standard ground joints, amber

50 ml, Amber, 51 mm, 29/32 NS
Catalog # 201-0926
Supplier:   Lenz Laborglas
Erlenmeyer flasks with standard ground joints, amber
Catalog # 201-0926
Supplier:  Lenz Laborglas
Erlenmeyer flasks with standard ground joints, amber
Erlenmeyer flasks with standard ground joints, amber

50 ml, Amber, 51 mm, 29/32 NS

Price
64,20 €
Log in for availability
1 * 1 items
Product Details & Documents

DURAN® borosilicate glass, brown.

  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes

Coloured brown to protect light-sensitive substances. Volume graduations and labelling area.

Documents

Specifications
  • Ext.Ø:
    51 mm
  • Socket size:
    29/32 NS
  • Capacity:
    50 ml
  • Colour:
    Amber
  • Seq:
    04
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.