Orders LinkContactUsLinkComponent
Biopsy processing/embedding cassettes, Microsette™ M503
Grey
Catalog # 720-0967
Supplier:   Simport Scientific
Biopsy processing/embedding cassettes, Microsette™ M503
Catalog # 720-0967
Supplier:  Simport Scientific
Biopsy processing/embedding cassettes, Microsette™ M503
Biopsy processing/embedding cassettes, Microsette™ M503
Grey
Price
524,00 €
Log in for availability
1 * 1.000 items
Product Details & Documents
Acetal polymer.
  • No need for biopsy pads
  • Six compartments, each 7×12 mm, numbered 1 to 6
  • 2000 square openings (each 0,26 mm) to maximise fluid exchange
  • Integral lid
  • Anterior writing area 45° angle suitable for some labelling systems
Specifications
  • Colour:
    Grey