Orders LinkContactUsLinkComponent
Biopsy processing/embedding cassettes, Micromesh™ M508
Salmon
Catalog # 720-0987
Supplier:   Simport Scientific
Biopsy processing/embedding cassettes, Micromesh™ M508
Catalog # 720-0987
Supplier:  Simport Scientific
Biopsy processing/embedding cassettes, Micromesh™ M508
Biopsy processing/embedding cassettes, Micromesh™ M508
Salmon
Price
518,00 €
Log in for availability
1 * 1.000 items
Product Details & Documents
Acetal polymer.
  • No need for biopsy pads
  • Cassettes have four square compartments each 13 mm
  • Cover and base have about 1676 square openings to maximise fluid exchange
  • Integral lid
  • Anterior writing area 45° angle suitable for some labelling systems
Specifications
  • Colour:
    Salmon