Orders LinkContactUsLinkComponent
Biopsy processing/embedding cassettes, Micromesh™ M507
Biopsy processing/embedding cassettes, Micromesh™ M507
Grey
Catalog # 720-0978
Supplier:   Simport Scientific
Biopsy processing/embedding cassettes, Micromesh™ M507
Catalog # 720-0978
Supplier:  Simport Scientific
Biopsy processing/embedding cassettes, Micromesh™ M507
Biopsy processing/embedding cassettes, Micromesh™ M507
Grey
Price
518,00 €
Log in for availability
1 * 1.000 items
Product Details & Documents
Acetal polymer. Biopsy processing/embedding cassettes, Micromesh™ M507.
  • No need for biopsy pads
  • Large square compartment, 27 mm², suitable for needle biopsies
  • 1676 square openings (0,38 mm²) allow improved fluid exchange
  • Integral lid
  • Anterior writing area 45° angle suitable for some labelling systems
Documents
Specifications
  • Colour:
    Grey