Orders LinkContactUsLinkComponent
Embedding rings
Embedding rings
Blue
Catalog # 720-0819
Supplier:   Simport Scientific
Embedding rings
Catalog # 720-0819
Supplier:  Simport Scientific
Embedding rings
Embedding rings
Blue
Price
151,00 €
Log in for availability
1 * 1.000 items
Product Details & Documents
PS. These embedding rings are suitable for holding and identifying tissue sample blocks and fit well in microtome chuck adapters.
  • The etched writing surface on the ring is marked with an identification number and placed on top of the sample block
  • Additional paraffin is poured into the base mould to cement the ring onto the tissue block
  • The embedding ring securely holds the tissue sample in the microtome chuck adapter for sectioning and then identifies the sample while in storage
Specifications
  • Colour:
    Blue