Orders LinkContactUsLinkComponent
Embedding rings
Yellow
  720-0818
Embedding rings
  720-0818
Embedding rings
Embedding rings
Yellow
151,00 €
1 * 1.000 items
PS. These embedding rings are suitable for holding and identifying tissue sample blocks and fit well in microtome chuck adapters.
  • The etched writing surface on the ring is marked with an identification number and placed on top of the sample block
  • Additional paraffin is poured into the base mould to cement the ring onto the tissue block
  • The embedding ring securely holds the tissue sample in the microtome chuck adapter for sectioning and then identifies the sample while in storage
  • Colour:
    Yellow