Orders LinkContactUsLinkComponent
VWR®, Slide Boxes
Slide boxes, White, PP, 100, 165×210×35 mm
  631-0735
VWR®, Slide Boxes
  631-0735
VWR®, Slide Boxes
VWR®, Slide Boxes
Slide boxes, White, PP, 100, 165×210×35 mm
12,50 €
1 * 1 items
Slide boxes for holding standard microscopy slides.
  • Hinged lids held securely by a latch lock
  • Stacking feet maximise stability when placed on top of one another
  • Slide inventory sheet on the inside of the lid
W×D×H: 90×140×35 mm (for 25), 165×210×35 mm (for 100)
  • No. of slides:
    100
  • Colour:
    White
  • Description:
    Slide boxes
  • Material:
    PP
  • Seq:
    002
  • W×D×H:
    165×210×35 mm