Replacement Cables
Replacement cable. Connect an Agilent 5890 GC to a non-Agilent integrator or standard strip chart recorder, 1,8 m
Catalog # REST20652
Supplier: Restek
Jump to:
Product Details & Documents
These are replacement cables for Agilent GCs, integrators and autosamplers.
Delivery information: Instructions and writing diagrams included.
Specifications
- Description:Replacement cable. Connect an Agilent 5890 GC to a non-Agilent integrator or standard strip chart recorder
- Length:1,8 m
- Seq:01