Internal reducing union, zero dead volume, Valco®
Internal reducing union, Stainless steel, 1,6 mm
REST20153
: Restek
Internal reducing union used to connect two sizes of tubing, using zero-dead-volume fittings on each end. Suitable for 1,6 to 3,2 mm OD tubing.
: Supplied with stainless steel ferrules.
- Hole Ø:1,6 mm
- Description:Internal reducing union
- Material:Stainless steel
- Seq:03