Order Entry
Slovenia
Orders LinkContactUsLinkComponent
Internal reducing union, zero dead volume, Valco®

Internal reducing union, zero dead volume, Valco®

Internal reducing union, Stainless steel, 0,75 mm
Catalog # REST20152
Supplier:   Restek
Internal reducing union, zero dead volume, Valco®
Catalog # REST20152
Supplier:  Restek
Internal reducing union, zero dead volume, Valco®
Internal reducing union, zero dead volume, Valco®

Internal reducing union, Stainless steel, 0,75 mm

Price
118,89 €
Log in for availability
1 * 1 items
Product Details & Documents

Internal reducing union used to connect two sizes of tubing, using zero-dead-volume fittings on each end. Suitable for 1,6 to 3,2 mm OD tubing.

Delivery information: Supplied with stainless steel ferrules.

Specifications
  • Hole Ø:
    0,75 mm
  • Description:
    Internal reducing union
  • Material:
    Stainless steel
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.