Orders LinkContactUsLinkComponent
Containers, rectangular
Containers, rectangular
23 l, 460×330×205 mm, White, HDPE
Catalog # SEMA1297
Supplier:   SEMADENI
Containers, rectangular
Catalog # SEMA1297
Supplier:  SEMADENI
Containers, rectangular
23 l, 460×330×205 mm, White, HDPE
Price
48,20 €
Log in for availability
1 * 1 items
Product Details & Documents
HDPE or PP, white, stackable.
These are ideal containers with side handles and edge for gripping to handle solid samples. Containers over 225 mm height are not suitable for liquids.
Specifications
  • Capacity:
    23 l
  • Colour:
    White
  • Material:
    HDPE
  • Seq:
    04
  • W×D×H:
    460×330×205 mm