Adhesive plasters, Salvequick
Salvequick plastic plasters refill, incl. 45 sterile plasters (27 pcs 72×19 mm and 18 pcs 72×25 mm)
Catalog # 118-0026
Supplier: ORKLA CARE
Jump to:
Product Details & Documents
These plaster refills are available in different versions: Made from plastic or fabric, blue detectable and for sensitive skin.
- In combination with dispenser: Complete, easy to use system
- Plasters are always available, ready to use
- Plasters are allergy-tested
- Each adhesive bandage is individually wrapped and sterile
Detectable plasters are specially designed for use in the food industry. The plasters are detectable by most metal detectors used within the food industry. The blue colour also makes the plasters easy to see.
Sensitive plasters are white and made of polyurethane. They are water repellent and suitable for sensitive skin.
Specifications
- Description:Salvequick plastic plasters refill, incl. 45 sterile plasters (27 pcs 72×19 mm and 18 pcs 72×25 mm)
- Seq:25