Orders LinkContactUsLinkComponent
Acetone
Acetone
5 L
Catalog # ROTH9372.6
Flammable
Irritant
Supplier:   Roth Carl
Acetone
Catalog # ROTH9372.6
Supplier:  Roth Carl
CAS Number:  67-64-1
Acetone
CAS Number:  67-64-1
Price
120,00 €
Log in for availability
1 * 5 L
Product Details & Documents
Synonyms:  2-Propanone, DMK, Propanone, Dimethyl ketone
  • CAS Number:67-64-1
  • Formula:CH₃COCH₃
  • MDL Number:MFCD00008765
  • Molecular Weight:58.08 g/mol
  • Merck Index:13,00067
  • Boiling Pt:56.2 °C (1013 hPa)
  • Melting Pt:–95.4 °C
  • Density:0.792 g/cm³ (20 °C)
  • Flash Pt:<–20 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:200-662-2
  • UN:1090
  • ADR:3,II
  • REACH:01-2119471330-49
Specifications
Specifications
  • Pk:
    5 L
Specifications