Order Entry
Slovenia
Orders LinkContactUsLinkComponent
Disodium hydrogen phosphate

Disodium hydrogen phosphate

Glass bottle for solids, 1 kg
Catalog # ROTHP030.2
Supplier:   Roth Carl
Disodium hydrogen phosphate
Catalog # ROTHP030.2
Supplier:  Roth Carl
CAS Number:  7558-79-4
Disodium hydrogen phosphate

Glass bottle for solids, 1 kg

CAS Number:  7558-79-4
Price
119,00 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  sec-Sodium phosphate, di-Sodium hydrogen orthophosphate, di-Sodium hydrogen phosphate anhydrous, Sodium hydrogen phosphate, Disodium phosphate, Sodium phosphate dibasic
  • CAS Number:7558-79-4
  • Formula:Na₂HPO₄
  • MDL Number:MFCD00003496
  • Molecular Weight:141.96 g/mol
  • Merck Index:12,08805
  • Melting Pt:250 °C
  • Density:1.53 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-448-7
  • REACH:01-2119489797-11
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications