N,N-Dimethylformamide
Glass bottle, 2,5 L
Catalog # ROTHA529.1
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Formic acid dimethylamide, Formdimethylamide, Dimethylformamide, DMF
- CAS Number:68-12-2
- Formula:HCON(CH₃)₂
- MDL Number:MFCD00003284
- Molecular Weight:73.09 g/mol
- Merck Index:13,03269
- Boiling Pt:153 °C (1013 hPa)
- Melting Pt:–61 °C
- Density:0.949 g/cm³ (20 °C)
- Flash Pt:57.5 °C (closed cup)
- Storage Temperature:Ambient
- EINECS:200-679-5
- UN:2265
- ADR:3,III
- REACH:01-2119475605-32
Specifications
Specifications
- Pack type:Glass bottle
- Pk:2,5 L



