Orders LinkContactUsLinkComponent
Water
Water
30 L
Catalog # ROTH3175.2
Supplier:   Roth Carl
Water
Catalog # ROTH3175.2
Supplier:  Roth Carl
CAS Number:  7732-18-5
CAS Number:  7732-18-5
Price
56,00 €
Log in for availability
1 * 30 L
Product Details & Documents
Synonyms:  Hydrogen oxide
  • CAS Number:7732-18-5
  • Formula:H₂O
  • MDL Number:MFCD00011332
  • Molecular Weight:18.02 g/mol
  • Merck Index:13,10098
  • Boiling Pt:100 °C (1013 hPa)
  • Melting Pt:0 °C
  • Density:1 g/cm³ (4 °C)
  • Storage Temperature:Ambient
  • EINECS:231-791-2
Specifications
Specifications
  • Pk:
    30 L