Order Entry
Slovenia
Orders LinkContactUsLinkComponent
2-Chlorophenol ≥99%

2-Chlorophenol ≥99%

Glass bottle for solids, 250 g
Catalog # A10311.30
Irritant
Environment
2-Chlorophenol ≥99%
Catalog # A10311.30
CAS Number:  95-57-8
2-Chlorophenol ≥99%
2-Chlorophenol ≥99%

Glass bottle for solids, 250 g

CAS Number:  95-57-8
Price
51,90 €
Log in for availability
1 * 250 g
Product Details & Documents

Purity: 99% CAS No.: 95-57-8 Molecular Formula: C6H5ClO Formula Weight: 128.56 Flash Point: 63°(145°F) Refractive Index: 1.5590 Odour: Phenol-like Melting Point: 7-8° Boiling Point: 175-176° Beilstein Reference: 1905114 Merck Reference: 14,2154 UN No.: 2021 MDL No.: MFCD00002159

Synonyms:  1-Chloro-2-hydroxybenzene
  • CAS Number:95-57-8
  • Formula:C₆H₅ClO
  • MDL Number:MFCD00002159
  • Molecular Weight:128.56 g/mol
  • Merck Index:14,2154
  • Boiling Pt:175…176 °C
  • Melting Pt:7…8 °C
  • Density:1.260
  • Flash Pt:63 °C (145 °F)
  • Storage Temperature:Ambient
  • EINECS:202-433-2
  • UN:2021
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.