Orders LinkContactUsLinkComponent
Micro haematocrit capillaries, VITREX®
VITREX® Capillaries, natrium-heparinized (red), 75 µl (10×100 pieces), 75 mm, 1,55 mm, 1,15
  720-2680
Micro haematocrit capillaries, VITREX®
  720-2680
Micro haematocrit capillaries, VITREX®
Micro haematocrit capillaries, VITREX®
VITREX® Capillaries, natrium-heparinized (red), 75 µl (10×100 pieces), 75 mm, 1,55 mm, 1,15
5,70 €
1 * 100 items
These tubes are supplied fire polished at the colour-coded end, which should be sealed with the Sigillum Wax. Fire polishing reduces wear on the centrifuge seals.
  • Each heparinised tube contains at least 4 IU heparin as prescribed by the Standard
  • Tubes are available in two types of packaging: Dispenser vials of 100 tubes each for hygienic and easy handling; and economy cardboard boxes, which reduce transport and handling costs
  • Sigillum Wax plate (needs to be ordered separately) has 2×24 numbered positions per pair, which makes it easy to seal and transport samples
  • The even wax layer ensures uniform sealing
 : According to DIN ISO 12772.
  • Ext.Ø:
    1,55 mm
  • Ø int.:
    1,15
  • Description:
    VITREX® Capillaries, natrium-heparinized (red), 75 µl (10×100 pieces)
  • Length:
    75 mm
  • Seq:
    13
Accessories
  •  
    Description
  • MODU140013
    Sigillum wax plate, with 24 numbered positions, 1,16×86 mm (Ø×L)
    1 * 10 items - 54,30 €