KIT CHEMETS OXYGEN DISSOLVED 5-180 PPB 1 * 1 Each
Catalog # CHETK-7518
Supplier: CHEMetrics
Frequently Bought Together
2 components: 100mg/l each of Rb Pr in HNO3 5% 1 * 50 mL
Supplier: CUSTOM MADE CHEMICALS LAB
Reagents for DEHA determination with Lovibond® MD photometers
Supplier: Lovibond
Fuchsin (acid) for tissue, stain, Sigma-Aldrich®
Supplier: Merck
ImmunoHistoMount™ Mounting Medium
Supplier: SIGMA ALDRICH MICROSCOPY
Reagents for ammonia determination using Lovibond® MD photometers
Supplier: Lovibond
SULFITES 10 MG/L IN H2O 1 * 100 mL
Supplier: CUSTOM MADE CHEMICALS LAB
FerroZine Powder, 500 g 1 * 500 g
Supplier: Hach
NEOMYCIN ELISA KIT 1 * 1 KIT
Supplier: ENZO LIFE SCIENCES
1 COMPONENT OS 1000MG/L IN HCL 2% 1 * 100 mL
Supplier: CUSTOM MADE CHEMICALS LAB
13 COMPONENTS: 1000MG/L EACH 1 * 5 Ampoul
Supplier: CUSTOM MADE CHEMICALS LAB
Decalcifier, rapid, histograde, for diagnostics use 1 * 2,5 L
Supplier: Avantor