Molecular models: covalent crystal structures
Molecular model of sodium chloride
Catalog # 764-0068
Supplier: SPIRING ENTERPRISES
Jump to:
Product Details & Documents
Structures of ionic crystals, sodium chloride.
- 36 atoms
- 80 bonds: 18 metal (grey), 18 halogen (green), 80 medium grey bonds
36 atoms.
80 bonds: 18 metal (grey), 18 halogen (green), 80 medium grey bonds.
Specifications
- Description:Molecular model of sodium chloride