Orders LinkContactUsLinkComponent
Clamps
Clamps
38 - 60 mm
Catalog # 241-0372
Supplier:   USBECK Laborgeräte
Clamps
Catalog # 241-0372
Clamps
Clamps
38 - 60 mm
Price
17,50 €
Log in for availability
1 * 1 items
Product Details & Documents
Zinc die-casting, nickel plated.
  • With vinyl coated jaws
  • With rod Ø 10 mm
Specifications
  • Spanning width:
    38 - 60 mm