Orders LinkContactUsLinkComponent
Clamps
Clamps
12 - 25 mm
Catalog # 241-0374
Supplier:   USBECK Laborgeräte
Clamps
Catalog # 241-0374
Clamps
Clamps
12 - 25 mm
Price
15,30 €
Log in for availability
1 * 1 items
Product Details & Documents
Zinc die-casting, nickel plated.
  • With vinyl coated jaws
  • With rod Ø 10 mm
Specifications
  • Spanning width:
    12 - 25 mm