Orders LinkContactUsLinkComponent
Dropping pipettes
Dropping pipettes
Dropping pipette, glass, with German inscription "Normal 20 Tropfen = 1 g", without rubber teat, 85 mm
Catalog # 612-3759
Dropping pipettes
Catalog # 612-3759
Dropping pipettes
Dropping pipettes
Dropping pipette, glass, with German inscription "Normal 20 Tropfen = 1 g", without rubber teat, 85 mm
Price
4,10 €
Log in for availability
1 * 1 items
Product Details & Documents
Dropping pipettes made of clear glass. Available in different styles.
Specifications
  • Description:
    Dropping pipette, glass, with German inscription "Normal 20 Tropfen = 1 g", without rubber teat
  • Length:
    85 mm