ContactUsLinkComponent
Cylinder graduated, capacity 5ml,hexagonal base with pourout, diameter 12.9mm, height 115mm, high form, white graduation, class b, g raduation divisions 0.1mm. 1 * 1 items
Catalog # 612-3769
Supplier:  KAVALIER GLASS
Cylinder graduated, capacity 5ml,hexagonal base with pourout, diameter 12.9mm, height 115mm, high form, white graduation, class b, g raduation divisions 0.1mm. 1 * 1 items
Catalog # 612-3769
Supplier:  KAVALIER GLASS
Frequently Bought Together