ContactUsLinkComponent
Cylinder graduated, capacity 100ml,hexagonal base with pourout, diameter 41mm, height 170mm, low form, white graduation, class b, gr aduation divisions 2.0mm. 1 * 1 items
Catalog # 612-3780
Supplier:  KAVALIER GLASS
undefined
Cylinder graduated, capacity 100ml,hexagonal base with pourout, diameter 41mm, height 170mm, low form, white graduation, class b, gr aduation divisions 2.0mm. 1 * 1 items
Catalog # 612-3780
Supplier:  KAVALIER GLASS
Frequently Bought Together