Microscope slides, SuperFrost®, Menzel Gläser
Yellow, Cellophane wrapped box
Catalog # 630-2017
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Microslide Superfrost 45° yellow glass cello ISO 8037/1 1,0mm 1 * 50 items Catalog #MENZAB00008332E01
Product MENZAB00008332E01 is a replacement for 630-2017 and has the same packaging and specifications, but may be a different quantity.
Microslide Superfrost 45° yellow glass with box cello ISO 8037/1 1,0mm 1 * 2.000 items Catalog #MENZAB00008332E042
Product MENZAB00008332E042 is a replacement for 630-2017 and has the same packaging and specifications, but may be a different quantity.
Product Details & Documents
These slides are made of extra-white glass, washed clean and polished.
- Labelling area resistant to all common solvents
- Six colours for identification and colour coding
- Thick labelling area to prevent slides from sticking together or getting scratched
Suitable for in vitro diagnostic (IVD) applications.
Complies with ISO 8037/I
Certifications: ISO 8037/I.
Packaging: In cardboard boxes of 50.
Documents
Specifications
- Packed:Cellophane wrapped box
- Colour:Yellow
- Seq:214