Microscope slides, SuperFrost®, Menzel Gläser
Pink, Cellophane wrapped box
630-1989
These slides are made of extra-white glass, washed clean and polished.
- Labelling area resistant to all common solvents
- Six colours for identification and colour coding
- Thick labelling area to prevent slides from sticking together or getting scratched
Suitable for in vitro diagnostic (IVD) applications.
Complies with ISO 8037/I
: ISO 8037/I.
: In cardboard boxes of 50.
- Packed:Cellophane wrapped box
- Colour:Pink
- Seq:14